ADVH49B8375E AChemBlock

(2-(morpholinomethyl)phenyl)methanol

  • Cas Number : 91271-63-5
  • Molecular Weight : 207.27

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin China
Product Link https://www.achemblock.com/2-morpholinomethyl-phenyl-methanol.html

Safety Information

MSDS https://www.achemblock.com/AC_SDS.php?Catalog=S89525
Availability details for
Quantity
  • Availability
  • Add to Cart