ADVH49B83762 AChemBlock

N,N-dimethylpyridin-4-amine 4-methylbenzenesulfonate

  • Cas Number : 91944-64-8
  • Molecular Weight : 294.37

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin China
Product Link https://www.achemblock.com/n-n-dimethylpyridin-4-amine-4-methylbenzenesulfonate.html

Safety Information

MSDS https://www.achemblock.com/AC_SDS.php?Catalog=S89529
Availability details for
Quantity
  • Availability
  • Add to Cart