ADVH49B83764 AChemBlock

ethyl 2-chloro-4-iodobenzoate

  • Cas Number : 92712-69-1
  • Molecular Weight : 310.52

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin China
Product Link https://www.achemblock.com/ethyl-2-chloro-4-iodobenzoate.html

Safety Information

MSDS https://www.achemblock.com/AC_SDS.php?Catalog=S89531
Availability details for
Quantity
  • Availability
  • Add to Cart