ADVH49B83766 AChemBlock

methyl 3-(aminomethyl)benzoate

  • Cas Number : 93071-65-9
  • Molecular Weight : 165.19

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin China
Product Link https://www.achemblock.com/methyl-3-aminomethyl-benzoate.html

Safety Information

MSDS https://www.achemblock.com/AC_SDS.php?Catalog=S89534
Availability details for
Quantity
  • Availability
  • Add to Cart