ADVH49B8376C AChemBlock

3-bromo-4-ethylbenzoic acid

  • Cas Number : 99548-53-5
  • Molecular Weight : 229.07

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin China
Product Link https://www.achemblock.com/3-bromo-4-ethylbenzoic-acid.html

Safety Information

MSDS https://www.achemblock.com/AC_SDS.php?Catalog=S89540
Availability details for
Quantity
  • Availability
  • Add to Cart