ADVH49B8376D AChemBlock

2-phenoxyphenol

  • Molecular Weight : 186.21

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin China
Product Link https://www.achemblock.com/2-phenoxyphenol.html

Safety Information

MSDS https://www.achemblock.com/AC_SDS.php?Catalog=S89541
Availability details for
Quantity
  • Availability
  • Add to Cart