ADVH49B83773 AChemBlock

4-methoxybenzene-1,3-diol

  • Cas Number : 6100-60-3
  • Molecular Weight : 140.14

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link https://www.achemblock.com/4-methoxybenzene-1-3-diol.html

Safety Information

MSDS https://www.achemblock.com/AC_SDS.php?Catalog=S89547
Availability details for
Quantity
  • Availability
  • Add to Cart