ADVH49B83774 AChemBlock

2-(4-bromophenyl)-1-(2,4-dihydroxyphenyl)ethan-1-one

  • Cas Number : 92152-60-8
  • Molecular Weight : 307.14

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link https://www.achemblock.com/2-4-bromophenyl-1-2-4-dihydroxyphenyl-ethan-1-one.html

Safety Information

MSDS https://www.achemblock.com/AC_SDS.php?Catalog=S89548
Availability details for
Quantity
  • Availability
  • Add to Cart