ADVH49B83779 AChemBlock

1,4,4-trimethyl-6-nitro-3,4-dihydroquinolin-2(1H)-one

  • Cas Number : 144583-89-1
  • Molecular Weight : 234.26

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link https://www.achemblock.com/1-4-4-trimethyl-6-nitro-3-4-dihydroquinolin-2-1h-one.html

Safety Information

MSDS https://www.achemblock.com/AC_SDS.php?Catalog=S89557
Availability details for
Quantity
  • Availability
  • Add to Cart