ADVH49B8377A AChemBlock

5-(pyrrolidin-1-yl)pyrimidine-2,4(1H,3H)-dione

  • Cas Number : 37454-54-9
  • Molecular Weight : 181.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Country of Origin China
Product Link https://www.achemblock.com/5-pyrrolidin-1-yl-pyrimidine-2-4-1h-3h-dione.html

Safety Information

MSDS https://www.achemblock.com/AC_SDS.php?Catalog=S89567
Availability details for
Quantity
  • Availability
  • Add to Cart