ADVH49B8377B AChemBlock

2-(bromomethyl)-1-methoxy-4-nitrobenzene

  • Cas Number : 3913-23-3
  • Molecular Weight : 246.06

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link https://www.achemblock.com/2-bromomethyl-1-methoxy-4-nitrobenzene.html

Safety Information

MSDS https://www.achemblock.com/AC_SDS.php?Catalog=S89568
Availability details for
Quantity
  • Availability
  • Add to Cart