ADVH49B8377C AChemBlock

CHF 5074

  • Cas Number : 749269-83-8
  • Molecular Weight : 325.16

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link https://www.achemblock.com/chf-5074.html

Safety Information

MSDS https://www.achemblock.com/AC_SDS.php?Catalog=S89577
Availability details for
Quantity
  • Availability
  • Add to Cart