ADVH49B83781 AChemBlock

5-bromo-2-methyl-4-nitropyridine

  • Cas Number : 1378873-37-0
  • Molecular Weight : 217.02

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link https://www.achemblock.com/5-bromo-2-methyl-4-nitropyridine.html

Safety Information

MSDS https://www.achemblock.com/AC_SDS.php?Catalog=S89587
Availability details for
Quantity
  • Availability
  • Add to Cart