ADVH49B83784 AChemBlock

1-(3-methoxy-2-nitrophenyl)ethan-1-one

  • Cas Number : 33852-43-6
  • Molecular Weight : 195.17

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link https://www.achemblock.com/1-3-methoxy-2-nitrophenyl-ethan-1-one.html

Safety Information

MSDS https://www.achemblock.com/AC_SDS.php?Catalog=S89590
Availability details for
Quantity
  • Availability
  • Add to Cart