ADVH49B83785 AChemBlock

ethyl 4-hydroxy-5-phenyl-1H-pyrazole-3-carboxylate

  • Cas Number : 34035-06-8
  • Molecular Weight : 232.24

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link https://www.achemblock.com/ethyl-4-hydroxy-5-phenyl-1h-pyrazole-3-carboxylate.html

Safety Information

MSDS https://www.achemblock.com/AC_SDS.php?Catalog=S89591
Availability details for
Quantity
  • Availability
  • Add to Cart