ADVH49B83786 AChemBlock

1,2-diphenyl-1,2,4-triazolidine-3,5-dione

  • Cas Number : 34877-12-8
  • Molecular Weight : 253.26

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link https://www.achemblock.com/1-2-diphenyl-1-2-4-triazolidine-3-5-dione.html

Safety Information

MSDS https://www.achemblock.com/AC_SDS.php?Catalog=S89592
Availability details for
Quantity
  • Availability
  • Add to Cart