ADVH49B8378C AChemBlock

Tubastatin A trifluoroacetate

  • Cas Number : 1239262-52-2
  • Molecular Weight : 337.42

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Country of Origin China
Product Link https://www.achemblock.com/tubastatin-a-trifluoroacetate.html

Safety Information

MSDS https://www.achemblock.com/AC_SDS.php?Catalog=S89598
Availability details for
Quantity
  • Availability
  • Add to Cart