ADVH49B8378D AChemBlock

MK-571

  • Cas Number : 135028-15-8
  • Molecular Weight : 515.08

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Country of Origin China
Product Link https://www.achemblock.com/mk-571.html

Safety Information

MSDS https://www.achemblock.com/AC_SDS.php?Catalog=S89599
Availability details for
Quantity
  • Availability
  • Add to Cart