ADVH49B83791 AChemBlock

Antineoplaston A10

  • Cas Number : 77658-84-5
  • Molecular Weight : 246.27

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Country of Origin China
Product Link https://www.achemblock.com/antineoplaston-a10.html

Safety Information

MSDS https://www.achemblock.com/AC_SDS.php?Catalog=S89603
Availability details for
Quantity
  • Availability
  • Add to Cart