ADVH49B83793 AChemBlock

2-(bromomethyl)-3,5-dichloropyridine

  • Cas Number : 1227502-19-3
  • Molecular Weight : 240.91

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link https://www.achemblock.com/2-bromomethyl-3-5-dichloropyridine.html

Safety Information

MSDS https://www.achemblock.com/AC_SDS.php?Catalog=S89606
Availability details for
Quantity
  • Availability
  • Add to Cart