ADVH49B83794 AChemBlock

methyl 4-bromo-6-chloronicotinate

  • Cas Number : 1256790-93-8
  • Molecular Weight : 250.48

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link https://www.achemblock.com/methyl-4-bromo-6-chloronicotinate.html

Safety Information

MSDS https://www.achemblock.com/AC_SDS.php?Catalog=S89607
Availability details for
Quantity
  • Availability
  • Add to Cart