ADVH49B83795 AChemBlock

2-(1H-pyrazol-1-yl)acetohydrazide

  • Cas Number : 934175-49-2
  • Molecular Weight : 140.15

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link https://www.achemblock.com/2-1h-pyrazol-1-yl-acetohydrazide.html

Safety Information

MSDS https://www.achemblock.com/AC_SDS.php?Catalog=S89621
Availability details for
Quantity
  • Availability
  • Add to Cart