ADVH49B83798 AChemBlock

(S)-morpholine-2-carbonitrile hydrochloride

  • Cas Number : 1359658-57-3
  • Molecular Weight : 148.59

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link https://www.achemblock.com/s-morpholine-2-carbonitrile-hydrochloride-311500.html

Safety Information

MSDS https://www.achemblock.com/AC_SDS.php?Catalog=S88191
Availability details for
Quantity
  • Availability
  • Add to Cart