ADVH49B83799 AChemBlock

1-methyl-3-(trimethylsilyl)naphthalen-2-ol

  • Molecular Weight : 230.38

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link https://www.achemblock.com/1-methyl-3-trimethylsilyl-naphthalen-2-ol.html

Safety Information

MSDS https://www.achemblock.com/AC_SDS.php?Catalog=S88664
Availability details for
Quantity
  • Availability
  • Add to Cart