ADVH49B8379A AChemBlock

Boc-D-phe(CBZ-4-Amino)-OH

  • Molecular Weight : 414.46

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link https://www.achemblock.com/boc-d-phe-cbz-4-amino-oh.html

Safety Information

MSDS https://www.achemblock.com/AC_SDS.php?Catalog=S89629
Availability details for
Quantity
  • Availability
  • Add to Cart