ADVH49B8379B AChemBlock

Sildenafil Chlorosulfone Impurity

  • Cas Number : 139756-22-2
  • Molecular Weight : 410.88

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin China
Product Link https://www.achemblock.com/sildenafil-chlorosulfone-impurity.html

Safety Information

MSDS https://www.achemblock.com/AC_SDS.php?Catalog=S89630
Availability details for
Quantity
  • Availability
  • Add to Cart