ADVH49B8379D AChemBlock

CBZ-

  • Molecular Weight : 518.7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link https://www.achemblock.com/cbz-hoasp-otbu-oh-dcha.html

Safety Information

MSDS https://www.achemblock.com/AC_SDS.php?Catalog=S89637
Availability details for
Quantity
  • Availability
  • Add to Cart