ADVH49B8379E AChemBlock

1-(2,3-dihydro-1H-pyrrolo[2,3-b]pyridin-1-yl)propan-1-one

  • Molecular Weight : 176.22

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link https://www.achemblock.com/1-2-3-dihydro-1h-pyrrolo-2-3-b-pyridin-1-yl-propan-1-one.html

Safety Information

MSDS https://www.achemblock.com/AC_SDS.php?Catalog=S88665
Availability details for
Quantity
  • Availability
  • Add to Cart