ADVH49B8379F AChemBlock

Fmoc-D-

  • Molecular Weight : 397.48

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link https://www.achemblock.com/fmoc-d-homo-ser-otbu-oh.html

Safety Information

MSDS https://www.achemblock.com/AC_SDS.php?Catalog=S89644
Availability details for
Quantity
  • Availability
  • Add to Cart