ADVH49B837A1 AChemBlock

Cbz-H-L-beta-homo-Glu(OtBu)-OH

  • Molecular Weight : 351.4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Country of Origin China
Product Link https://www.achemblock.com/cbz-h-l-beta-homo-glu-otbu-oh.html

Safety Information

MSDS https://www.achemblock.com/AC_SDS.php?Catalog=S89646
Availability details for
Quantity
  • Availability
  • Add to Cart