ADVH49B837A2 AChemBlock

Boc-His(trt)-aib-OH

  • Cas Number : 2061897-68-3
  • Molecular Weight : 582.71

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Country of Origin China
Product Link https://www.achemblock.com/boc-his-trt-aib-oh.html

Safety Information

MSDS https://www.achemblock.com/AC_SDS.php?Catalog=S89647
Availability details for
Quantity
  • Availability
  • Add to Cart