ADVH49B837A4 AChemBlock

Cbz-MeAsp(OtBu)-OH

  • Cas Number : 42417-70-9
  • Molecular Weight : 337.38

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Country of Origin China
Product Link https://www.achemblock.com/cbz-measp-otbu-oh.html

Safety Information

MSDS https://www.achemblock.com/AC_SDS.php?Catalog=S89649
Availability details for
Quantity
  • Availability
  • Add to Cart