ADVH49B837A8 AChemBlock

CBZ-D-

  • Molecular Weight : 394.47

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link https://www.achemblock.com/cbz-d-holys-boc-oh.html

Safety Information

MSDS https://www.achemblock.com/AC_SDS.php?Catalog=S89660
Availability details for
Quantity
  • Availability
  • Add to Cart