ADVH49B837A9 AChemBlock

Boc-D-

  • Molecular Weight : 385.46

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link https://www.achemblock.com/boc-d-homo-tyr-obzl-oh.html

Safety Information

MSDS https://www.achemblock.com/AC_SDS.php?Catalog=S89661
Availability details for
Quantity
  • Availability
  • Add to Cart