ADVH49B837AD AChemBlock

(R)-3-((((9H-fluoren-9-yl)methoxy)carbonyl)amino)-4,4-diphenylbutanoic acid

  • Molecular Weight : 477.57

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link https://www.achemblock.com/r-3-9h-fluoren-9-yl-methoxy-carbonyl-amino-4-4-diphenylbutanoic-acid-313547.html

Safety Information

MSDS https://www.achemblock.com/AC_SDS.php?Catalog=S89668
Availability details for
Quantity
  • Availability
  • Add to Cart