ADVH49B837AF AChemBlock

D-

  • Molecular Weight : 231.68

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link https://www.achemblock.com/d-homo-tyr-oh-hydrochloride.html

Safety Information

MSDS https://www.achemblock.com/AC_SDS.php?Catalog=S89675
Availability details for
Quantity
  • Availability
  • Add to Cart