ADVH49B837B2 AChemBlock

(R)-3-amino-4,4-diphenylbutanoic acid hydrochloride

  • Molecular Weight : 291.78

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link https://www.achemblock.com/r-3-amino-4-4-diphenylbutanoic-acid-hydrochloride-313557.html

Safety Information

MSDS https://www.achemblock.com/AC_SDS.php?Catalog=S89678
Availability details for
Quantity
  • Availability
  • Add to Cart