ADVH49B837B8 AChemBlock

L-beta-HoLys(Boc)-OH

  • Molecular Weight : 260.34

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link https://www.achemblock.com/l-beta-holys-boc-oh.html

Safety Information

MSDS https://www.achemblock.com/AC_SDS.php?Catalog=S89685
Availability details for
Quantity
  • Availability
  • Add to Cart