ADVH49B837BB AChemBlock

Fmoc-D-beta-Homo-Trp(Boc)-OH

  • Molecular Weight : 540.62

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link https://www.achemblock.com/fmoc-d-beta-homo-trp-boc-oh.html

Safety Information

MSDS https://www.achemblock.com/AC_SDS.php?Catalog=S89688
Availability details for
Quantity
  • Availability
  • Add to Cart