ADVH49B837BE AChemBlock

Oct3 and 4-inducer-1

  • Cas Number : 1016535-83-3
  • Molecular Weight : 241.27

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link https://www.achemblock.com/oct3-and-4-inducer-1.html

Safety Information

MSDS https://www.achemblock.com/AC_SDS.php?Catalog=S89699
Availability details for
Quantity
  • Availability
  • Add to Cart