ADVH49B837BF AChemBlock

Duvoglustat Hydrochloride

  • Cas Number : 308362-25-6
  • Molecular Weight : 388.44

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin China
Product Link https://www.achemblock.com/duvoglustat-hydrochloride.html

Safety Information

MSDS https://www.achemblock.com/AC_SDS.php?Catalog=S89700
Availability details for
Quantity
  • Availability
  • Add to Cart