ADVH49B837C2 AChemBlock

1-hydroxyimino-1-(4-)propan-2-one

  • Cas Number : 36176-47-3
  • Molecular Weight : 193.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin China
Product Link https://www.achemblock.com/1-hydroxyimino-1-4-propan-2-one.html

Safety Information

MSDS https://www.achemblock.com/AC_SDS.php?Catalog=S89718
Availability details for
Quantity
  • Availability
  • Add to Cart