ADVH49B837C4 AChemBlock

NS3728

  • Cas Number : 265646-85-3
  • Molecular Weight : 495.18

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin China
Product Link https://www.achemblock.com/ns3728.html

Safety Information

MSDS https://www.achemblock.com/AC_SDS.php?Catalog=S89720
Availability details for
Quantity
  • Availability
  • Add to Cart