ADVH49B837C6 AChemBlock

Boc-Ser(Allyl)-OH

  • Cas Number : 150438-78-1
  • Molecular Weight : 245.28

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Country of Origin China
Product Link https://www.achemblock.com/boc-ser-allyl-oh.html

Safety Information

MSDS https://www.achemblock.com/AC_SDS.php?Catalog=S89725
Availability details for
Quantity
  • Availability
  • Add to Cart