ADVH49B837C7 AChemBlock

tert-butyl 7-benzyl-9-hydroxy-9-(trifluoromethyl)-3,7-diaza-bicyclo[3 3 1]nonane-3-carboxylate (A)

  • Molecular Weight : 400.44

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link https://www.achemblock.com/tert-butyl-7-benzyl-9-hydroxy-9-trifluoromethyl-3-7-diaza-bicyclo-3-3-1-nonane-3-carboxylate-a.html

Safety Information

MSDS https://www.achemblock.com/AC_SDS.php?Catalog=S89726
Availability details for
Quantity
  • Availability
  • Add to Cart