ADVH49B837C9 AChemBlock

tert-butyl 4-hydroxy-4-(3-hydroxypropyl)piperidine-1-carboxylate

  • Cas Number : 374794-88-4
  • Molecular Weight : 259.35

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link https://www.achemblock.com/tert-butyl-4-hydroxy-4-3-hydroxypropyl-piperidine-1-carboxylate.html

Safety Information

MSDS https://www.achemblock.com/AC_SDS.php?Catalog=S89728
Availability details for
Quantity
  • Availability
  • Add to Cart