ADVH49B837CF AChemBlock

Darusentan

  • Cas Number : 171714-84-4
  • Molecular Weight : 410.43

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link https://www.achemblock.com/darusentan.html

Safety Information

MSDS https://www.achemblock.com/AC_SDS.php?Catalog=S89741
Availability details for
Quantity
  • Availability
  • Add to Cart