ADVH49B837D2 AChemBlock

CBL-0137 hydrochloride

  • Cas Number : 1197397-89-9
  • Molecular Weight : 370.88

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Country of Origin China
Product Link https://www.achemblock.com/cbl-0137-hydrochloride.html

Safety Information

MSDS https://www.achemblock.com/AC_SDS.php?Catalog=S89746
Availability details for
Quantity
  • Availability
  • Add to Cart