ADVH49B837D6 AChemBlock

JNJ-38877605

  • Cas Number : 943540-74-7
  • Molecular Weight : 374.36

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin China
Product Link https://www.achemblock.com/jnj-38877605.html

Safety Information

MSDS https://www.achemblock.com/AC_SDS.php?Catalog=S89752
Availability details for
Quantity
  • Availability
  • Add to Cart