ADVH49B837DB AChemBlock

JNJ-10229570

  • Cas Number : 524923-88-4
  • Molecular Weight : 389.48

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link https://www.achemblock.com/jnj-10229570.html

Safety Information

MSDS https://www.achemblock.com/AC_SDS.php?Catalog=S89762
Availability details for
Quantity
  • Availability
  • Add to Cart